N-Acetyl-5-brom-2-(trifluormethoxy)anilin

Lieferant: COMBI-BLOCKS

HC-2835-5G HC-2835-1G
COBBHC-2835-5GEA 303 EUR
COBBHC-2835-5G COBBHC-2835-1G
N-Acetyl-5-brom-2-(trifluormethoxy)anilin
N-Acetyl-5-brom-2-(trifluormethoxy)anilin
Formel: C₉H₇BrF₃NO₂
MDL: MFCD02317366
CAS-Nummer: 392726-69-1

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR