3-Chlor-5-isopropoxyphenylboronsäurepinakolester

Lieferant: COMBI-BLOCKS

Synonyme: 2-(3-Chloro-5-isopropoxyphenyl)-4,4,5,5-tetramethyl-1,3,2-dioxaborolane

PN-5956-1G PN-5956-250M
COBBPN-5956-1GEA 983 EUR
COBBPN-5956-1G COBBPN-5956-250M
3-Chlor-5-isopropoxyphenylboronsäurepinakolester
3-Chlor-5-isopropoxyphenylboronsäurepinakolester
Formel: C₁₅H₂₂BClO₃
Molecular Weight: 296,6 g/mol
MDL: MFCD12546618
CAS-Nummer: 1218789-42-4

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR