3-Chlor-5-methoxyphenylboronsäurepinakolester

Lieferant: COMBI-BLOCKS

Synonyme: 2-(3-Chloro-5-methoxyphenyl)-4,4,5,5-tetramethyl-1,3,2-dioxaborolane

PN-2589-5G PN-2589-1G
COBBPN-2589-5GEA 1180 EUR
COBBPN-2589-5G COBBPN-2589-1G
3-Chlor-5-methoxyphenylboronsäurepinakolester
3-Chlor-5-methoxyphenylboronsäurepinakolester
Formel: C₁₃H₁₈BClO₃
Molecular Weight: 268,5442 g/mol
MDL: MFCD12405332
CAS-Nummer: 929626-16-4

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR