3-Chlor-5-propoxyphenylboronsäurepinakolester

Lieferant: COMBI-BLOCKS

Synonyme: 2-(3-Chloro-5-propoxyphenyl)-4,4,5,5-tetramethyl-1,3,2-dioxaborolane

PN-5955-1G PN-5955-250M
COBBPN-5955-1GEA 595 EUR
COBBPN-5955-1G COBBPN-5955-250M
3-Chlor-5-propoxyphenylboronsäurepinakolester
3-Chlor-5-propoxyphenylboronsäurepinakolester
Formel: C₁₅H₂₂BClO₃
Molecular Weight: 296.6000 g/mol
MDL: MFCD12546578
CAS-Nummer: 1218789-41-3

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR