2-Fluor-1,3-dimethoxy-4-nitrobenzol

Lieferant: COMBI-BLOCKS

Synonyme: 2,4-Dimethoxy-3-fluoronitrobenzene

AN-1118-5G AN-1118-1G
COBBAN-1118-5GEA 60.3 EUR
COBBAN-1118-5G COBBAN-1118-1G
2-Fluor-1,3-dimethoxy-4-nitrobenzol
2-Fluor-1,3-dimethoxy-4-nitrobenzol
Formel: C₈H₈FNO₄
Molecular Weight: 201,1500 g/mol
MDL: MFCD09027197
CAS-Nummer: 155020-44-3

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR