2-Chlor-5-(hydroxymethyl)phenylboronsäure

Lieferant: COMBI-BLOCKS

Synonyme: 2-Chloro-5-hydroxymethylphenylboronic acid

BB-9040-5G BB-9040-1G
COBBBB-9040-5GEA 312 EUR
COBBBB-9040-5G COBBBB-9040-1G
2-Chlor-5-(hydroxymethyl)phenylboronsäure
2-Chlor-5-(hydroxymethyl)phenylboronsäure
Formel: C₇H₈BClO₃
Molecular Weight: 186.4000 g/mol
MDL: MFCD06656270
CAS-Nummer: 1003042-59-8

Bestellen

TECHNISCHE DATEN

Environmentally Preferable

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR