3,4-Dichlor-2-methylphenylboronsäure

Lieferant: COMBI-BLOCKS

Synonyme: (3,4-Dichloro-2-methylphenyl)boronic acid

BB-3981-1G BB-3981-250M
COBBBB-3981-1GEA 798 EUR
COBBBB-3981-1G COBBBB-3981-250M
3,4-Dichlor-2-methylphenylboronsäure
3,4-Dichlor-2-methylphenylboronsäure
Formel: C₇H₇BCl₂O₂
Molecular Weight: 204.85 g/mol
MDL: MFCD09475873
CAS-Nummer: 957035-17-5

Bestellen

TECHNISCHE DATEN

Environmentally Preferable

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR