4-(Benzyloxy)-3-methylanilin Hydrochlorid

Lieferant: COMBI-BLOCKS

AN-1379-1G AN-1379-5G
COBBAN-1379-1GEA 149 EUR
COBBAN-1379-1G COBBAN-1379-5G
4-(Benzyloxy)-3-methylanilin Hydrochlorid
4-(Benzyloxy)-3-methylanilin Hydrochlorid
Formel: C₁₄H₁₆ClNO
MDL: MFCD12025952
CAS-Nummer: 1150114-24-1

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR