4-(3-Bromphenyl)morpholin

Lieferant: COMBI-BLOCKS

AN-1483-5G AN-1483-1G AN-1483-25G
COBBAN-1483-5GEA 130 EUR
COBBAN-1483-5G COBBAN-1483-1G COBBAN-1483-25G
4-(3-Bromphenyl)morpholin
4-(3-Bromphenyl)morpholin
Formel: C₁₀H₁₂BrNO
Molecular Weight: 242,1100 g/mol
MDL: MFCD04112585
CAS-Nummer: 197846-82-5

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR