4-(2-Fluor-4-nitrophenyl)morpholin
Lieferant: COMBI-BLOCKS
AN-1199-5G
AN-1199-1G
AN-1199-25G
COBBAN-1199-5GEA
31.2
EUR
COBBAN-1199-5G
COBBAN-1199-1G
COBBAN-1199-25G
4-(2-Fluor-4-nitrophenyl)morpholin
4-(2-Fluor-4-nitrophenyl)morpholin
Formel:
C₁₀H₁₁FN₂O₃ Molecular Weight: 226,20 g/mol |
MDL:
MFCD03138381 CAS-Nummer: 2689-39-6 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...