1-(Toluol-4-sulfonyl)indol-3-boronsäurepinakolester
Lieferant: COMBI-BLOCKS
Synonyme:
3-(4,4,5,5-Tetramethyl-1,3,2-dioxaborolan-2-yl)-1-tosyl-1H-indole
PN-8729-1G
PN-8729-5G
COBBPN-8729-1GEA
0
EUR
COBBPN-8729-1G
COBBPN-8729-5G
1-(Toluol-4-sulfonyl)indol-3-boronsäurepinakolester
1-(Toluol-4-sulfonyl)indol-3-boronsäurepinakolester
Formel:
C₂₁H₂₄BNO₄S |
MDL:
MFCD08063118 CAS-Nummer: 1073354-51-4 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...