1-(Toluol-4-sulfonyl)indol-3-boronsäure
Lieferant: COMBI-BLOCKS
BB-8729-5G
BB-8729-1G
COBBBB-8729-5GEA
1350
EUR
COBBBB-8729-5G
COBBBB-8729-1G
1-(Toluol-4-sulfonyl)indol-3-boronsäure
1-(Toluol-4-sulfonyl)indol-3-boronsäure
Formel:
C₁₅H₁₄BNO₄S Molecular Weight: 315,15 g/mol |
MDL:
MFCD03093101 CAS-Nummer: 149108-61-2 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...