4-(tert-Butoxycarbonyl)phenylboronsäurepinakolester

Lieferant: COMBI-BLOCKS

Synonyme: tert-Butyl 4-(4,4,5,5-Tetramethyl-1,3,2-dioxaborolan-2-yl)benzoate, 4-(tert-Butoxycarbonyl)phenylboronic acid, pinacol ester, 4-Boc-phenylboronic acid Pinacol ester, 2-(4-tert-Butoxycarbonylphenyl)-4,4,5,5-tetramethyl-1,3,2-dioxaborolane, 4-(4,4,5,5-Tetramethyl-1,3,2-dioxaborolan-2-yl)benzoic acid tert-Butyl ester, 4-(tert-Butoxycarbonyl)phenylboronic acid Pinacol ester

BB-2396-5G BB-2396-25G BB-2396-1G
COBBBB-2396-5GEA 131 EUR
COBBBB-2396-5G COBBBB-2396-25G COBBBB-2396-1G
4-(tert-Butoxycarbonyl)phenylboronsäurepinakolester
4-(tert-Butoxycarbonyl)phenylboronsäurepinakolester
Formel: C₁₇H₂₅BO₄
Molecular Weight: 304,1890 g/mol
MDL: MFCD03453055
CAS-Nummer: 850568-72-8

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR