3-(tert-Butoxycarbonyl)phenylboronsäure

Lieferant: COMBI-BLOCKS

Synonyme: 3-(tert-Butoxycarbonyl)benzeneboronic acid, 3-tert-Butoxycarbonylphenylboronic acid

BB-2119-5G BB-2119-25G BB-2119-1G
COBBBB-2119-5GEA 130 EUR
COBBBB-2119-5G COBBBB-2119-25G COBBBB-2119-1G
3-(tert-Butoxycarbonyl)phenylboronsäure
3-(tert-Butoxycarbonyl)phenylboronsäure
Formel: C₁₁H₁₅BO₄
Molecular Weight: 222,05 g/mol
MDL: MFCD01630855
CAS-Nummer: 220210-56-0

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR