3-(Isopropoxycarbonyl)phenylboronsäure

Lieferant: Thermo Scientific

Synonyme: 3-(Isopropoxycarbonyl)benzeneboronic acid

AC35950.250 AC35950.1
MAYBAC35950.250EA 0 EUR
MAYBAC35950.250 MAYBAC35950.1
3-(Isopropoxycarbonyl)phenylboronsäure
3-(Isopropoxycarbonyl)phenylboronsäure
Formel: C₁₀H₁₃BO₄
Molecular Weight: 208,02 g/mol
MDL: MFCD02093047
CAS-Nummer: 342002-80-6

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR