3-(Isopropoxycarbonyl)phenylboronsäure

Lieferant: COMBI-BLOCKS

Synonyme: 3-(Isopropoxycarbonyl)benzeneboronic acid

BB-2246-5G BB-2246-1G
COBBBB-2246-5GEA 34.6 EUR
COBBBB-2246-5G COBBBB-2246-1G
3-(Isopropoxycarbonyl)phenylboronsäure
3-(Isopropoxycarbonyl)phenylboronsäure
Formel: C₁₀H₁₃BO₄
Molecular Weight: 208,02 g/mol
MDL: MFCD02093047
CAS-Nummer: 342002-80-6

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR