5-Chlor-N,2-dimethylanilin Hydrochlorid
Lieferant: COMBI-BLOCKS
Synonyme:
N-Methyl 5-chloro-2-methylaniline hydrochloride
HC-2859-5G
HC-2859-1G
COBBHC-2859-5GEA
884
EUR
COBBHC-2859-5G
COBBHC-2859-1G
5-Chlor-N,2-dimethylanilin Hydrochlorid
5-Chlor-N,2-dimethylanilin Hydrochlorid
Formel:
C₈H₁₁Cl₂N |
MDL:
MFCD12756441 CAS-Nummer: 1187386-18-0 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...