4-(Methylthio)benzylalkohol ≥98%

Lieferant: Thermo Fisher Scientific

Synonyme: 4-(Methylthio)benzyl Alcohol

B22455.06 B22455.14 B22455.22 AB132566.0100
B22455.06EA 50.4 EUR
B22455.06 B22455.14 B22455.22 ABCRAB132566.0100
4-(Methylthio)benzylalkohol ≥98%
4-(Methylthio)benzylalkohol

Purity: 98% CAS No.: 3446-90-0 Molecular Formula: C8H10OS Formula Weight: 154.23 Flash Point: 110°(230°F) Melting Point: 41-43° Boiling Point: 108-109°/0.4mm MDL No.: MFCD00009706

Formel: C₈H₁₀OS
Molecular Weight: 154,23 g/mol
Siedepunkt: 108…109 °C (0,4 mmHg)
Schmelzpunkt: 41…43 °C
Flammpunkt: >110 °C (230 °F)
Lagertemperatur: Raumtemperatur
MDL: MFCD00009706
CAS-Nummer: 3446-90-0

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR