3-Chlor-2,6-xylidin ≥99%

Lieferant: Thermo Scientific
Gefahr

Synonyme: 3-Chloro-2,6-dimethylaniline

B22232.09 B22232.18 B22232.30
B22232.09EA 39.3 EUR
B22232.09 B22232.18 B22232.30
3-Chlor-2,6-xylidin ≥99%
3-Chlor-2,6-xylidin

Purity: 99% CAS No.: 26829-77-6 Molecular Formula: C8H10ClN Formula Weight: 155.63 Melting Point: 30-32° Boiling Point: 136-138°/17mm MDL No.: MFCD02683086

Formel: C₈H₁₀ClN
Molecular Weight: 155,63 g/mol
Siedepunkt: 136…138 °C (17 mmHg)
Schmelzpunkt: 30…32 °C
Lagertemperatur: Raumtemperatur
MDL: MFCD02683086
CAS-Nummer: 26829-77-6

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR