N-Formyl-L-methionin

Lieferant: BACHEM BIOCHEMICA

Synonyme: For-Met-OH, Formyl-L-methionine

4001407.0005 4001407.0025
BACE4001407.0005EA 0 EUR
BACE4001407.0005 BACE4001407.0025
N-Formyl-L-methionin
N-Formyl-L-methionin
Formel: C₆H₁₁NO₃S
Molecular Weight: 177.2 g/mol
MDL: MFCD00021033
CAS-Nummer: 4289-98-9

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR