N-Formyl-L-methionin, Sigma-Aldrich®

Lieferant: Merck

Synonyme: For-Met-OH, Formyl-L-methionine

M-Clarity™ Qualitätsstufe = MQ200

F3377-10MG F3377-1G F3377-250MG
SIALF3377-10MGEA 37.4 EUR
SIALF3377-10MG SIALF3377-1G SIALF3377-250MG
N-Formyl-L-methionin, Sigma-Aldrich®
N-Formyl-L-methionin
Formel: C₆H₁₁NO₃S
Molecular Weight: 177.2 g/mol
MDL: MFCD00021033
CAS-Nummer: 4289-98-9

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR