L-Methioninsulfon

Lieferant: BACHEM BIOCHEMICA

BACE4017827.0005EA 80.4 EUR
BACE4017827.0005 BACE4017827.0025
L-Methioninsulfon
L-Methioninsulfon
Formel: C₅H₁₁NO₄S
Molecular Weight: 181,21 g/mol
MDL: MFCD00066020
CAS-Nummer: 7314-32-1
EINECS: 230-774-7

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR