N-Cbz-D-Alaninol 98%

Lieferant: Thermo Scientific

Synonyme: (2R)-N-Benzyloxycarbonylamino-1-propanol, N-Cbz-D-Alaninol, Z-D-Alaninol, (R)-2-(Cbz-amino)-1-propanol

B22372.03 B22372.06 B22372.14
B22372.03EA 20.3 EUR
B22372.03 B22372.06 B22372.14
N-Cbz-D-Alaninol 98%
N-Cbz-D-Alaninol

Purity: 98% CAS No.: 61425-27-2 Molecular Formula: C11H15NO3 Formula Weight: 209.25 Storage Temperature: 2°C to 8°C Melting Point: 82-84° MDL No.: MFCD00672531

Formel: C₁₁H₁₅NO₃
Molecular Weight: 209,2 g/mol
Schmelzpunkt: 82…84 °C
MDL: MFCD00672531
CAS-Nummer: 61425-27-2

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR