N-Cbz-D-Alaninol, Sigma-Aldrich®

Lieferant: Merck

Synonyme: (2R)-N-Benzyloxycarbonylamino-1-propanol, N-Cbz-D-Alaninol, Z-D-Alaninol, (R)-2-(Cbz-amino)-1-propanol

536474-1G 536474-5G
SIAL536474-1GEA 47.9 EUR
SIAL536474-1G SIAL536474-5G
N-Cbz-D-Alaninol, Sigma-Aldrich®
N-Cbz-D-Alaninol
Formel: C₁₁H₁₅NO₃
Molecular Weight: 209,2 g/mol
MDL: MFCD00672531
CAS-Nummer: 61425-27-2

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR