Bromkresolpurpur 1 g/l in Methanol

Lieferant: Bernd Kraft

Synonyme: 5′,5''-Dibrom-o-kresolsulfonphthalein

20725.0700 20725.1700
KRAF20725.0700EA 65.4 EUR
KRAF20725.0700 KRAF20725.1700
Bromkresolpurpur 1 g/l in Methanol
Bromkresolpurpur
Formel: C₂₁H₁₆Br₂O₅S
Molecular Weight: 540.23 g/mol
Schmelzpunkt: 242
MDL: MFCD00011681
CAS-Nummer: 115-40-2
EINECS: 204-087-8
Merck Index: 13,01371

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR