1-Acetylpiperidin-4-carbonsäure ≥98%
Lieferant: Thermo Scientific
|
Achtung
|
A15606.06
A15606.14
A15606.22
A15606.06EA
22
EUR
A15606.06
A15606.14
A15606.22
1-Acetylpiperidin-4-carbonsäure ≥98%
1-Acetylpiperidin-4-carbonsäure
Purity: 98+%
CAS No.: 25503-90-6
Molecular Formula: C8H13NO3
Formula Weight: 171.19
Melting Point: 180-184°
Beilstein Reference: 389577
Formel:
C₈H₁₃NO₃ Molecular Weight: 171,19 g/mol Schmelzpunkt: 180…184 °C Lagertemperatur: Raumtemperatur |
MDL:
MFCD00023706 CAS-Nummer: 25503-90-6 EINECS: 247-049-6 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...