1-Acetylpiperidin-4-carbonsäure

Lieferant: FLUOROCHEM

002491.5G 002491-100G 002491-25G
FLUO002491.5GEA 28.7 EUR
FLUO002491.5G FLUO002491-100G FLUO002491-25G
1-Acetylpiperidin-4-carbonsäure
1-Acetylpiperidin-4-carbonsäure
Formel: C₈H₁₃NO₃
Molecular Weight: 171,19 g/mol
MDL: MFCD00023706
CAS-Nummer: 25503-90-6
EINECS: 247-049-6

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR