1-Acetylpiperidin-4-carbonsäure, Sigma-Aldrich®

Lieferant: Merck


M-Clarity™ Qualitätsstufe = MQ100

647756-25G 647756-5G
SIAL647756-25GEA 88.1 EUR
SIAL647756-25G SIAL647756-5G
1-Acetylpiperidin-4-carbonsäure, Sigma-Aldrich®
1-Acetylpiperidin-4-carbonsäure
Formel: C₈H₁₃NO₃
Molecular Weight: 171,19 g/mol
MDL: MFCD00023706
CAS-Nummer: 25503-90-6
EINECS: 247-049-6

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR