N(α)-Acetyl-L-Tryptophan, Sigma-Aldrich®
Lieferant: Merck
M-Clarity™ Qualitätsstufe = MQ200
A6376-10G
A6376-25G
SIALA6376-10GEA
73.6
EUR
SIALA6376-10G
SIALA6376-25G
N(α)-Acetyl-L-Tryptophan, Sigma-Aldrich®
N(α)-Acetyl-L-tryptophan
Formel:
C₁₃H₁₄N₂O₃ |
MDL:
MFCD00065976 CAS-Nummer: 1218-34-4 EINECS: 214-935-9 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...