1-(2-Brombenzoyl)piperidin

Lieferant: COMBI-BLOCKS

CA-4733-5G CA-4733-25G CA-4733-1G
COBBCA-4733-5GEA 646 EUR
COBBCA-4733-5G COBBCA-4733-25G COBBCA-4733-1G
1-(2-Brombenzoyl)piperidin
1-(2-Brombenzoyl)piperidin
Formel: C₁₂H₁₄BrNO
MDL: MFCD00449518
CAS-Nummer: 61153-35-3

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR