2-(3-Bromphenyl)-1-(pyrrolidin-1-yl)ethanon
Lieferant: COMBI-BLOCKS
CA-4454-5G
CA-4454-1G
COBBCA-4454-5GEA
1040
EUR
COBBCA-4454-5G
COBBCA-4454-1G
2-(3-Bromphenyl)-1-(pyrrolidin-1-yl)ethanon
2-(3-Bromphenyl)-1-(pyrrolidin-1-yl)ethanon
Formel:
C₁₂H₁₄BrNO |
MDL:
MFCD09800919 CAS-Nummer: 951884-73-4 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...