1-(3-Brombenzoyl)piperidin

Lieferant: COMBI-BLOCKS

CA-4493-5G CA-4493-1G
COBBCA-4493-5GEA 236 EUR
COBBCA-4493-5G COBBCA-4493-1G
1-(3-Brombenzoyl)piperidin
1-(3-Brombenzoyl)piperidin
Formel: C₁₂H₁₄BrNO
MDL: MFCD00194116
CAS-Nummer: 59507-53-8

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR