1-(3-Brombenzoyl)pyrrolidin
Lieferant: COMBI-BLOCKS
CA-4549-5G
CA-4549-25G
CA-4549-1G
COBBCA-4549-5GEA
303
EUR
COBBCA-4549-5G
COBBCA-4549-25G
COBBCA-4549-1G
1-(3-Brombenzoyl)pyrrolidin
1-(3-Brombenzoyl)pyrrolidin
Formel:
C₁₁H₁₂BrNO |
MDL:
MFCD00592553 CAS-Nummer: 346721-91-3 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...