1-(4-Brom-3-methylbenzoyl)pyrrolidin

Lieferant: COMBI-BLOCKS

Synonyme: (4-Bromo-3-methylphenylcarbonyl)pyrrolidine

CA-4559-5G CA-4559-1G
COBBCA-4559-5GEA 451 EUR
COBBCA-4559-5G COBBCA-4559-1G
1-(4-Brom-3-methylbenzoyl)pyrrolidin
1-(4-Brom-3-methylbenzoyl)pyrrolidin
Formel: C₁₂H₁₄BrNO
MDL: MFCD09972125
CAS-Nummer: 149105-15-7

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR