2-(Cyclopropylamino)-5-pyrimidinboronsäurepinakolester

Lieferant: COMBI-BLOCKS

Synonyme: N-Cyclopropyl-5-(4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl)pyrimidin-2-amine

PN-5930-1G PN-5930-250M
COBBPN-5930-1GEA 836 EUR
COBBPN-5930-1G COBBPN-5930-250M
2-(Cyclopropylamino)-5-pyrimidinboronsäurepinakolester
2-(Cyclopropylamino)-5-pyrimidinboronsäurepinakolester
Formel: C₁₃H₂₀BN₃O₂
Molecular Weight: 261,1300 g/mol
MDL: MFCD07375178
CAS-Nummer: 1218789-33-3

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR