4-Methoxy-2-methylanilin Hydrochlorid
Lieferant: COMBI-BLOCKS
AN-1372-5G
AN-1372-1G
COBBAN-1372-5GEA
451
EUR
COBBAN-1372-5G
COBBAN-1372-1G
4-Methoxy-2-methylanilin Hydrochlorid
4-Methoxy-2-methylanilin Hydrochlorid
Formel:
C₈H₁₂ClNO Molecular Weight: 173.6400 g/mol |
MDL:
MFCD00155304 CAS-Nummer: 133985-88-3 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...