2-Brom-N-(4-methoxybenzyl)-6-nitroanilin
Lieferant: COMBI-BLOCKS
AN-2556-5G
AN-2556-1G
COBBAN-2556-5GEA
1490
EUR
COBBAN-2556-5G
COBBAN-2556-1G
2-Brom-N-(4-methoxybenzyl)-6-nitroanilin
2-Brom-N-(4-methoxybenzyl)-6-nitroanilin
Formel:
C₁₄H₁₃BrN₂O₃ |
MDL:
MFCD15143452 CAS-Nummer: 1215205-09-6 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...