3-Brom-N-(4-methoxybenzyl)-2-nitroanilin
Lieferant: COMBI-BLOCKS
AN-1427-1G
AN-1427-5G
COBBAN-1427-1GEA
204
EUR
COBBAN-1427-1G
COBBAN-1427-5G
3-Brom-N-(4-methoxybenzyl)-2-nitroanilin
3-Brom-N-(4-methoxybenzyl)-2-nitroanilin
Formel:
C₁₄H₁₃BrN₂O₃ |
MDL:
MFCD11855828 CAS-Nummer: 1133115-36-2 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...