3-Chlor-4-(2'-chlorbenzyloxy)phenylboronsäure

Lieferant: COMBI-BLOCKS

Synonyme: (3-Chloro-4-((2-chlorobenzyl)oxy)phenyl)boronic acid

BB-8147-1G BB-8147-250M
COBBBB-8147-1GEA 215 EUR
COBBBB-8147-1G COBBBB-8147-250M
3-Chlor-4-(2'-chlorbenzyloxy)phenylboronsäure
3-Chlor-4-(2'-chlorbenzyloxy)phenylboronsäure
Formel: C₁₃H₁₁BCl₂O₃
Molecular Weight: 296,9400 g/mol
MDL: MFCD06798081
CAS-Nummer: 870777-26-7

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR