3-Chlor-4-(3'-chlorbenzyloxy)phenylboronsäure

Lieferant: COMBI-BLOCKS

Synonyme: (3-Chloro-4-((3-chlorobenzyl)oxy)phenyl)boronic acid

BB-8223-1G BB-8223-250M
COBBBB-8223-1GEA 243 EUR
COBBBB-8223-1G COBBBB-8223-250M
3-Chlor-4-(3'-chlorbenzyloxy)phenylboronsäure
3-Chlor-4-(3'-chlorbenzyloxy)phenylboronsäure
Formel: C₁₃H₁₁BCl₂O₃
Molecular Weight: 296,94 g/mol
MDL: MFCD06411318
CAS-Nummer: 849062-26-6

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR