3-Chlor-4-(2'-fluorbenzyloxy)phenylboronsäure
Lieferant: COMBI-BLOCKS
Synonyme:
(3-Chloro-4-((2-fluorobenzyl)oxy)phenyl)boronic acid
BB-8175-1G
BB-8175-250M
COBBBB-8175-1GEA
125
EUR
COBBBB-8175-1G
COBBBB-8175-250M
3-Chlor-4-(2'-fluorbenzyloxy)phenylboronsäure
3-Chlor-4-(2'-fluorbenzyloxy)phenylboronsäure
Formel:
C₁₃H₁₁BClFO₃ Molecular Weight: 280.4900 g/mol |
MDL:
MFCD06798082 CAS-Nummer: 870777-28-9 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...