3-Chlor-4-(4'-fluorbenzyloxy)phenylboronsäure

Lieferant: COMBI-BLOCKS

Synonyme: (3-Chloro-4-((4-fluorobenzyl)oxy)phenyl)boronic acid

BB-8029-1G BB-8029-250M
COBBBB-8029-1GEA 258 EUR
COBBBB-8029-1G COBBBB-8029-250M
3-Chlor-4-(4'-fluorbenzyloxy)phenylboronsäure
3-Chlor-4-(4'-fluorbenzyloxy)phenylboronsäure
Formel: C₁₃H₁₁BClFO₃
Molecular Weight: 280,490 g/mol
MDL: MFCD06411358
CAS-Nummer: 849062-39-1

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR