5-Fluor-8-chinolinol

Lieferant: FLUOROCHEM

006239-1G 006239-2.5G
FLUO006239-1GEA 713 EUR
FLUO006239-1G FLUO006239-2.5G
5-Fluor-8-chinolinol
5-Fluor-8-chinolinol
Formel: C₉H₆FNO
MDL: MFCD00059754
CAS-Nummer: 387-97-3

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR