1-Bromcyclohexen

Lieferant: COMBI-BLOCKS

Synonyme: 1-Bromo-1-cyclohexene, 1-Bromocyclohex-1-ene

OT-0350-1G OT-0350-5G
COBBOT-0350-1GEA 303 EUR
COBBOT-0350-1G COBBOT-0350-5G
1-Bromcyclohexen
1-Bromocyclohexene
Formel: C₆H₉Br
Molecular Weight: 161,0397 g/mol
MDL: MFCD09878413
CAS-Nummer: 2044-08-8

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR