4-Hydroxytetrahydropyran

Lieferant: COMBI-BLOCKS

Synonyme: Tetrahydro-2H-pyran-4-ol, Tetrahydro-4H-pyran-4-ol

AM-1701-5G AM-1701-1G
COBBAM-1701-5GEA 63.4 EUR
COBBAM-1701-5G COBBAM-1701-1G
4-Hydroxytetrahydropyran
4-Hydroxytetrahydropyran
Formel: C₅H₁₀O₂
Molecular Weight: 102,1317 g/mol
MDL: MFCD00006633
CAS-Nummer: 2081-44-9
EINECS: 218-210-8

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR