4-Hydroxytetrahydropyran, Sigma-Aldrich®

Lieferant: Merck

Synonyme: Tetrahydro-2H-pyran-4-ol, Tetrahydro-4H-pyran-4-ol

M-Clarity™ Qualitätsstufe = MQ200

198234-1G 198234-5G
SIAL198234-1GEA 26 EUR
SIAL198234-1G SIAL198234-5G
4-Hydroxytetrahydropyran, Sigma-Aldrich®
4-Hydroxytetrahydropyran
Formel: C₅H₁₀O₂
Molecular Weight: 102,1317 g/mol
MDL: MFCD00006633
CAS-Nummer: 2081-44-9
EINECS: 218-210-8

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR