6-Acetamidopicolinsäure
Lieferant: COMBI-BLOCKS
Synonyme:
6-Acetamidopyridine-2-carboxylic acid
CA-4142-5G
CA-4142-1G
COBBCA-4142-5GEA
471
EUR
COBBCA-4142-5G
COBBCA-4142-1G
6-Acetamidopicolinsäure
6-Acetamidopicolinsäure
Formel:
C₈H₈N₂O₃ Molecular Weight: 180,2 g/mol |
MDL:
MFCD00233707 CAS-Nummer: 26893-72-1 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...