4-Methoxypicolinamid

Lieferant: COMBI-BLOCKS

Synonyme: 4-Methoxypyridine-2-carboxamide

CA-4150-1G CA-4150-5G
COBBCA-4150-1GEA 141 EUR
COBBCA-4150-1G COBBCA-4150-5G
4-Methoxypicolinamid
4-Methoxypicolinamid
Formel: C₇H₈N₂O
Molecular Weight: 152,1506 g/mol
MDL: MFCD00955999
CAS-Nummer: 90151-10-3

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR