5-Methoxypicolinamid

Lieferant: COMBI-BLOCKS

PY-8048-1G PY-8048-5G PY-8048-250M
COBBPY-8048-1GEA 2030 EUR
COBBPY-8048-1G COBBPY-8048-5G COBBPY-8048-250M
5-Methoxypicolinamid
5-Methoxypicolinamid
Formel: C₇H₈N₂O
MDL: MFCD11976465
CAS-Nummer: 88166-65-8

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR