2-Brom-4'-(4-morpholino)acetophenon

Lieferant: COMBI-BLOCKS

Synonyme: 2-Bromo-1-[4-(morpholin-4-yl)phenyl]ethan-1-one

OT-0496-1G OT-0496-5G
COBBOT-0496-1GEA 286 EUR
COBBOT-0496-1G COBBOT-0496-5G
2-Brom-4'-(4-morpholino)acetophenon
2-Brom-4'-(4-morpholino)acetophenon
Formel: C₁₂H₁₄BrNO₂
MDL: MFCD03783555
CAS-Nummer: 210832-85-2

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR